Learn More
DWK Life Sciences Kimble™ KIMAX™ Brand Colorware Volumetric Class A Flasks
Single graduation ring blasted on the neck and durable matte finish marking area for use with an ordinary pencil
115.00 € - 569.00 €
Product Code | Brand | Capacity (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Color | Price | Quantity & Availability | ||||
10532483
|
DWK Life Sciences
28014R-25 |
25 mL | Raging Red |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10121582
|
DWK Life Sciences
28014B-25 |
25 mL | Bright Blue |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10445732
|
DWK Life Sciences
28014R-50 |
50 mL | Raging Red |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10089781
|
DWK Life Sciences
28014B-50 |
50 mL | Bright Blue |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10049761
|
DWK Life Sciences
28014Y-50 |
50 mL | Sunny Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10385262
|
DWK Life Sciences
28014R-100 |
100 mL | Raging Red |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10407462
|
DWK Life Sciences
28014Y-100 |
100 mL | Sunny Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10099661
|
DWK Life Sciences
28014Y-500 |
500 mL | Sunny Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10203352
|
DWK Life Sciences
28014R-500 |
500 mL | Raging Red |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10733293
|
DWK Life Sciences
28014B-500 |
500 mL | Bright Blue |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10049631
|
DWK Life Sciences
28014R-1000 |
1000 mL | Raging Red |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Flasks with brightly colored capacity and marking spot for easy reading and identification. Available in four colors. Matte-finish marking spot can be marked with an ordinary pencil. Calibrated “To Contain” with single graduation ring blasted on neck. Come with standard taper ground glass stopper. Designed from requirements for Class A “unserialized” flasks of ASTM E 288.
Specifications
Glass | |
Borosilicate Glass | |
Yes |