Learn More
DWK Life Sciences Kimble™ KIMAX™ Brand Colorware Volumetric Class A Flasks
Single graduation ring blasted on the neck and durable matte finish marking area for use with an ordinary pencil
115.00 € - 569.00 €
Specifications
Closure Material Glass
Material Borosilicate Glass
Graduated Yes
View More Specs
Products 11
Product Code Brand Capacity (Metric) Color Price Quantity & Availability  
Product Code Brand Capacity (Metric) Color Price Quantity & Availability  
10532483
View Documents Promotion Details
DWK Life Sciences
28014R-25
25 mL Raging Red This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10121582
View Documents Promotion Details
DWK Life Sciences
28014B-25
25 mL Bright Blue This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10445732
View Documents Promotion Details
DWK Life Sciences
28014R-50
50 mL Raging Red This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10089781
View Documents Promotion Details
DWK Life Sciences
28014B-50
50 mL Bright Blue This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10049761
View Documents Promotion Details
DWK Life Sciences
28014Y-50
50 mL Sunny Yellow This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10385262
View Documents Promotion Details
DWK Life Sciences
28014R-100
100 mL Raging Red This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10407462
View Documents Promotion Details
DWK Life Sciences
28014Y-100
100 mL Sunny Yellow This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10099661
View Documents Promotion Details
DWK Life Sciences
28014Y-500
500 mL Sunny Yellow This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10203352
View Documents Promotion Details
DWK Life Sciences
28014R-500
500 mL Raging Red This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10733293
View Documents Promotion Details
DWK Life Sciences
28014B-500
500 mL Bright Blue This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10049631
View Documents Promotion Details
DWK Life Sciences
28014R-1000
1000 mL Raging Red This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Flasks with brightly colored capacity and marking spot for easy reading and identification. Available in four colors. Matte-finish marking spot can be marked with an ordinary pencil. Calibrated “To Contain” with single graduation ring blasted on neck. Come with standard taper ground glass stopper. Designed from requirements for Class A “unserialized” flasks of ASTM E 288.
Specifications
Specifications
Glass
Borosilicate Glass