Learn More
DWK Life Sciences Kimble™ KIMAX™ Brand Colorware Volumetric Class A Flasks
Single graduation ring blasted on the neck and durable matte finish marking area for use with an ordinary pencil
Brand:  DWK Life Sciences 28014Y-500
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10099661
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Calibrated to contain with Class A accuracy but without serial number
  • Standard taper ground glass stopper is provided
  • Designed from ASTM Specification E288, Class A unserialized requirements
Specifications
Specifications
Glass
500 mL
11.29 in.
±0.2 mL
19
Sunny Yellow
Borosilicate Glass
28.7 cm