CAS RN 76272-56-5
CAS RN 76272-56-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
endo-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
endo-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
endo-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.