Learn More
endo-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine, TRC
Specifications
Specifications
| Chemical Name or Material | (1R,3r,5S)-9-methyl-9-azabicyclo(3.3.1)nonan-3-amine |
| CAS | 76272-56-5 |
| Molecular Formula | C9 H18 N2 |
| Quantity | 50 mg |
| Synonym | (1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]-nonan-3-amine, endo-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine, endo-3-Amino-9-methyl-9-azabicyclo[3.3.1]nonane, Granisetron Hydrochloride Imp. E (EP) |
| InChI Formula | InChI=1S/C9H18N2/c1-11-8-3-2-4-9(11)6-7(10)5-8/h7-9H,2-6,10H2,1H3/t7-,8+,9- |
| SMILES | CN1[C@@H]2CCC[C@H]1C[C@@H](N)C2 |
| IUPAC Name | (1R,5S)-9-methyl-9-azabicyclo[3.3.1]nonan-3-amine |
| Molecular Weight (g/mol) | 154.253 |
| Formula Weight | 154.147 |