Learn More
(1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine, Mikromol™
Description
(1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine
Specifications
Specifications
| Chemical Name or Material | (1R,3r,5S)-9-methyl-9-azabicyclo(3.3.1)nonan-3-amine |
| CAS | 76272-56-5 |
| Analyte or Component Names | (1R,3r,5S)-9-methyl-9-azabicyclo(3.3.1)nonan-3-amine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H18 N2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | (1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine, Granisetron Hydrochloride Imp. E (EP), Granisetron Imp. E (EP), Granisetron USP Related Compound E, Granisetron USP RC E, (1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]-nonan-3-amine, endo-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine, endo-3-Amino-9-methyl-9-azabicyclo[3.3.1]nonane, Granisetron Related Compound E, Granisetron Hydrochloride Impurity E, Granisetron Impurity E |
| InChI Formula | InChI=1S/C9H18N2/c1-11-8-3-2-4-9(11)6-7(10)5-8/h7-9H,2-6,10H2,1H3/t7-,8+,9- |
| SMILES | CN1[C@@H]2CCC[C@H]1C[C@@H](N)C2 |
| IUPAC Name | (1R,5S)-9-methyl-9-azabicyclo[3.3.1]nonan-3-amine |
| Show More |