CAS RN 584-03-2
CAS RN 584-03-2
IUPAC Name:
butane-1,2-diol
Synonyms:
1,2-butanediol α-butylene glycol α-butyleneglycol 1,2-butylene glycol ethylethylene glycol 1,2-(dihydroxy)butane
Molecular Weight (g/mol):
90.12
Molecular Formula:
C4H10O2
InChi Key:
BMRWNKZVCUKKSR-UHFFFAOYNA-N
SMILES:
CCC(O)CO
1
–
4
of
4
results
1
1,2-Butanediol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,2-Butanediol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,2-Butanediol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
4
1,2-Butanediol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.