Learn More
1,2-Butanediol, TRC
Specifications
Specifications
| Chemical Name or Material | 1,2-Butanediol |
| CAS | 584-03-2 |
| Molecular Formula | C4 H10 O2 |
| Quantity | 5 g |
| Synonym | (±)-Butane-1,2-diol, 1,2-Butylene glycol, 1,2-Dihydroxybutane, DL-1,2-Butanediol, NSC 24242 |
| InChI Formula | InChI=1S/C4H10O2/c1-2-4(6)3-5/h4-6H,2-3H2,1H3 |
| SMILES | CCC(O)CO |
| IUPAC Name | butane-1,2-diol |
| Molecular Weight (g/mol) | 90.12 |
| Formula Weight | 90.0681 |