Learn More
1,2-Butanediol, Dr. Ehrenstorfer
Product Code. 30327083
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327083 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30327083 Supplier Dr. Ehrenstorfer Supplier No. DRECA10861290
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,2-Butanediol
CAS 584-03-2
Analyte or Component Names 1,2-Butanediol
Shipping Condition +20°C
Molecular Formula C4 H10 O2
Certifications/Compliance ISO/IEC 17025
Synonym (±)-Butane-1,2-diol, 1,2-Butylene glycol, 1,2-Dihydroxybutane, DL-1,2-Butanediol, NSC 24242
InChI Formula InChI=1S/C4H10O2/c1-2-4(6)3-5/h4-6H,2-3H2,1H3
SMILES CCC(O)CO
IUPAC Name butane-1,2-diol
Molecular Weight (g/mol) 90.12
Quantity 250 mg
Formula Weight 90.0681 g/mol