CAS RN 53242-76-5
CAS RN 53242-76-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-[(4-Chlorophenyl)acetyl]benzoic Acid, TRC
CAS: 53242-76-5 Molecular Formula: C15 H11 Cl O3 Molecular Weight (g/mol): 274.7 Synonym: 2-[(4-Chlorophenyl)acetyl]benzoic Acid,Azelastine Hydrochloride Imp. C (EP) IUPAC Name: 2-[2-(4-chlorophenyl)acetyl]benzoic acid SMILES: OC(=O)c1ccccc1C(=O)Cc2ccc(Cl)cc2
2
2-[(4-Chlorophenyl)acetyl]benzoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.