Learn More
2-[(4-Chlorophenyl)acetyl]benzoic Acid, Mikromol™
Description
2-[(4-Chlorophenyl)acetyl]benzoic Acid
Specifications
Specifications
| Chemical Name or Material | 2-[(4-Chlorophenyl)acetyl]benzoic Acid |
| CAS | 53242-76-5 |
| Analyte or Component Names | 2-[(4-Chlorophenyl)acetyl]benzoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H11 Cl O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Azelastine Hydrochloride Imp. C (EP), Azelastine Imp. C (EP), 2-[(4-Chlorophenyl)acetyl]benzoic Acid, Azelastine Hydrochloride Impurity C, Azelastine Impurity C |
| InChI Formula | InChI=1S/C15H11ClO3/c16-11-7-5-10(6-8-11)9-14(17)12-3-1-2-4-13(12)15(18)19/h1-8H,9H2,(H,18,19) |
| SMILES | OC(=O)c1ccccc1C(=O)Cc2ccc(Cl)cc2 |
| IUPAC Name | 2-[2-(4-chlorophenyl)acetyl]benzoic acid |
| Show More |