Learn More
2-[(4-Chlorophenyl)acetyl]benzoic Acid, TRC
Product Code. 17953575
Change view
Click to view available options
Quantity:
1 g
2.5 g
10 g
Unit Size:
10g
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17953575 2.5 g 2.5g
17923475 1 g 1g
17904265 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17953575 Supplier Toronto Research Chemicals Supplier No. TRCC3754502.5G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 53242-76-5
Molecular Formula C15 H11 Cl O3
Molecular Weight (g/mol) 274.7
Synonym 2-[(4-Chlorophenyl)acetyl]benzoic Acid, Azelastine Hydrochloride Imp. C (EP)
IUPAC Name 2-[2-(4-chlorophenyl)acetyl]benzoic acid
SMILES OC(=O)c1ccccc1C(=O)Cc2ccc(Cl)cc2
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C15H11ClO3/c16-11-7-5-10(6-8-11)9-14(17)12-3-1-2-4-13(12)15(18)19/h1-8H,9H2,(H,18,19)
Formula Weight 274.0397
Percent Purity >95%
Chemical Name or Material 2-[(4-Chlorophenyl)acetyl]benzoic Acid
Grade HPLC