CAS RN 283594-90-1
CAS RN 283594-90-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Spiromesifen, TRC
CAS: 283594-90-1 Molecular Formula: C23 H30 O4 Molecular Weight (g/mol): 370.48 Synonym: Butanoic acid, 3,3-dimethyl-, 2-oxo-3-(2,4,6-trimethylphenyl)-1-oxaspiro[4.4]non-3-en-4-yl ester,BSN 2060,Forbid,Oberon,Spiromesifen IUPAC Name: [2-oxo-3-(2,4,6-trimethylphenyl)-1-oxaspiro[4.4]non-3-en-4-yl] 3,3-dimethylbutanoate SMILES: Cc1cc(C)c(C2=C(OC(=O)CC(C)(C)C)C3(CCCC3)OC2=O)c(C)c1
2
Spiromesifen 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality