Learn More
Spiromesifen 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30309034
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309034 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30309034 Supplier Dr. Ehrenstorfer Supplier No. DREA16972970AL100
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Spiromesifen
CAS 283594-90-1
Analyte or Component Names Spiromesifen
Shipping Condition +20°C
Molecular Formula C23 H30 O4
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Butanoic acid, 3,3-dimethyl-, 2-oxo-3-(2,4,6-trimethylphenyl)-1-oxaspiro[4.4]non-3-en-4-yl ester, BSN 2060, Forbid, Oberon, Spiromesifen
InChI Formula InChI=1S/C23H30O4/c1-14-11-15(2)18(16(3)12-14)19-20(26-17(24)13-22(4,5)6)23(27-21(19)25)9-7-8-10-23/h11-12H,7-10,13H2,1-6H3
SMILES Cc1cc(C)c(C2=C(OC(=O)CC(C)(C)C)C3(CCCC3)OC2=O)c(C)c1
IUPAC Name [2-oxo-3-(2,4,6-trimethylphenyl)-1-oxaspiro[4.4]non-3-en-4-yl] 3,3-dimethylbutanoate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 370.48
Quantity 1 mL
Formula Weight 370.2144 g/mol