Learn More
Spiromesifen, TRC
Product Code. 30136030
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136030 250 mg 250mg
17900545 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30136030 Supplier Toronto Research Chemicals Supplier No. TRCS683505250MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 283594-90-1
Molecular Formula C23 H30 O4
Molecular Weight (g/mol) 370.48
Synonym Butanoic acid, 3,3-dimethyl-, 2-oxo-3-(2,4,6-trimethylphenyl)-1-oxaspiro[4.4]non-3-en-4-yl ester, BSN 2060, Forbid, Oberon, Spiromesifen
IUPAC Name [2-oxo-3-(2,4,6-trimethylphenyl)-1-oxaspiro[4.4]non-3-en-4-yl] 3,3-dimethylbutanoate
SMILES Cc1cc(C)c(C2=C(OC(=O)CC(C)(C)C)C3(CCCC3)OC2=O)c(C)c1
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C23H30O4/c1-14-11-15(2)18(16(3)12-14)19-20(26-17(24)13-22(4,5)6)23(27-21(19)25)9-7-8-10-23/h11-12H,7-10,13H2,1-6H3
Formula Weight 370.2144
Chemical Name or Material Spiromesifen