CAS RN 151767-02-1
CAS RN 151767-02-1
IUPAC Name:
sodium 2-[1-({[(1R)-1-{3-[(1E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl}-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanyl}methyl)cyclopropyl]acetate
Synonyms:
singulair montelukast sodium salt singular montair montelukast sodium
Molecular Weight (g/mol):
608.17
Molecular Formula:
C35H35ClNNaO3S
InChi Key:
LBFBRXGCXUHRJY-HKHDRNBDSA-M
SMILES:
[Na+].CC(C)(O)C1=CC=CC=C1CC[C@@H](SCC1(CC([O-])=O)CC1)C1=CC=CC(\C=C\C2=CC=C3C=CC(Cl)=CC3=N2)=C1
1
–
6
of
6
results
1
Promotions Available
Montelukast sodium, 98%
Montelukast sodium, 98%
2
Montelukast Sodium, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Montelukast sodium, MedChemExpress
MedChemExpress Montelukast sodium (MK0476) is a potent, selective and orally active antagonist of cysteinyl leukotriene receptor 1 (CysLT1).
4
Tocris Bioscience™ Montelukast sodium
CysLT1 and GPR17 antagonist
5
6