Learn More
Montelukast sodium, MedChemExpress
Product Code. 16461807
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
Product Code. 16461807
Brand: Medchem Express HY13315
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Montelukast sodium (MK0476) is a potent, selective and orally active antagonist of cysteinyl leukotriene receptor 1 (CysLT1).
Montelukast sodium can be used for the reseach of asthma and liver injury. Montelukast sodium also has an antioxidant effect in intestinal ischemia-reperfusion injury, and could reduce cardiac damage. Montelukast sodium decreases eosinophil infiltration into the asthmatic airways. Montelukast sodium can also be used for COVID-19 research.
Specifications
Chemical Name or Material Montelukast sodium
Physical Form Solid
CAS 151767-02-1
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C35H35ClNNaO3S
For Use With (Application) COVID-19-immunoregulation
Synonym MK0476
Solubility Information DMSO : 50 mg/mL (82.21 mM; Need ultrasonic)
SMILES ClC1=CC2=C(C=C1)C=CC(/C=C/C3=CC([C@H](SCC4(CC(O[Na])=O)CC4)CCC5=CC=CC=C5C(C)(O)C)=CC=C3)=N2
Molecular Weight (g/mol) 608.17
Formula Weight 608.17
Percent Purity 98.62%
Grade Research