Learn More
Montelukast Sodium, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Montelukast Sodium
Specifications
Specifications
| Chemical Name or Material | Montelukast Sodium |
| CAS | 151767-02-1 |
| Analyte or Component Names | Montelukast Sodium |
| Shipping Condition | Room Temperature |
| Molecular Formula | C35 H35 Cl N O3 S . Na |
| Certifications/Compliance | ISO 17034 |
| Synonym | Montelukast Sodium, 1-[[[(1R)-1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]-cyclopropaneacetic acid monosodium salt, [R-(E)]-1-[[[1-[3-[2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]-cyclopropaneacetic acid monosodium salt, MK 476, Montelukast monosodium salt, Singulair, Sodium montelukast |
| InChI Formula | InChI=1S/C35H36ClNO3S.Na/c1-34(2,40)30-9-4-3-7-25(30)13-17-32(41-23-35(18-19-35)22-33(38)39)27-8-5-6-24(20-27)10-15-29-16-12-26-11-14-28(36)21-31(26)37-29;/h3-12,14-16,20-21,32,40H,13,17-19,22-23H2,1-2H3,(H,38,39);/q;+1/p-1/b15-10+;/t32-;/m1./s1 |
| SMILES | [Na+].CC(C)(O)c1ccccc1CC[C@@H](SCC2(CC(=O)[O-])CC2)c3cccc(\C=C\c4ccc5ccc(Cl)cc5n4)c3 |
| IUPAC Name | sodium;2-[1-[[(1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanylmethyl]cyclopropyl]acetate |
| Show More |