accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13182-89-3

CH3NNOONOO

CAS RN 13182-89-3

IUPAC Name: 2-(2-methyl-5-nitro-1H-imidazol-1-yl)ethyl benzoate
Synonyms: metrogyl metronidazole benzoate flegyl benzoylmetronidazole
Molecular Weight (g/mol): 275.26
Molecular Formula: C13H13N3O4
InChi Key: CUUCCLJJOWSASK-UHFFFAOYSA-N
SMILES: CC1=NC=C(N1CCOC(=O)C1=CC=CC=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results