Learn More
Metronidazole Benzoate, LoGiCal
Product Code. 30462362
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 30462362
Brand: LoGiCal LGCFOR0278.00
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Metronidazole Benzoate
CAS 13182-89-3
Analyte or Component Names Metronidazole Benzoate
Shipping Condition Room Temperature
Molecular Formula C13 H13 N3 O4
Certifications/Compliance ISO 9001
Synonym Metronidazole Benzoate, 2-Methyl-5-nitro-1H-imidazole-1-ethanol 1-benzoate, 2-Methyl-5-nitro-1H-imidazole-1-ethanol benzoate (ester), 2-Methyl-5-nitroimidazole-1-ethanol 1-benzoate, 2-Methyl-5-nitroimidazole-1-ethanol 1-benzoate (ester), Benzoylmetronidazole, Klion suspension, Metronidazole benzoate
InChI Formula InChI=1S/C13H13N3O4/c1-10-14-9-12(16(18)19)15(10)7-8-20-13(17)11-5-3-2-4-6-11/h2-6,9H,7-8H2,1H3
Solution Type Neat
SMILES O=C(OCCN1C(=NC=C1N(=O)=O)C)C=2C=CC=CC2
IUPAC Name 2-(2-methyl-5-nitroimidazol-1-yl)ethyl benzoate
Industry Type Clinical Testing
Molecular Weight (g/mol) 275.26
Quantity 10 mg
Formula Weight 275.0906 g/mol