Learn More
Metronidazole Benzoate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Metronidazole Benzoate
Specifications
Specifications
| Chemical Name or Material | Metronidazole Benzoate |
| CAS | 13182-89-3 |
| Analyte or Component Names | Metronidazole Benzoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H13 N3 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Metronidazole Benzoate, 2-Methyl-5-nitro-1H-imidazole-1-ethanol 1-benzoate, 2-Methyl-5-nitro-1H-imidazole-1-ethanol benzoate (ester), 2-Methyl-5-nitroimidazole-1-ethanol 1-benzoate, 2-Methyl-5-nitroimidazole-1-ethanol 1-benzoate (ester), Benzoylmetronidazole, Klion suspension, Metronidazole benzoate |
| InChI Formula | InChI=1S/C13H13N3O4/c1-10-14-9-12(16(18)19)15(10)7-8-20-13(17)11-5-3-2-4-6-11/h2-6,9H,7-8H2,1H3 |
| SMILES | Cc1ncc([N+](=O)[O-])n1CCOC(=O)c2ccccc2 |
| IUPAC Name | 2-(2-methyl-5-nitroimidazol-1-yl)ethyl benzoate |
| Show More |