CAS RN 842138-74-3
CAS RN 842138-74-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
5-Bromoquinazolin-6-ylthiourea, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-(5-Bromoquinoxalin-6-yl)thiourea, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
5-Bromoquinazolin-6-ylthiourea, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.