Learn More
1-(5-Bromoquinoxalin-6-yl)thiourea, Mikromol™
Description
1-(5-Bromoquinoxalin-6-yl)thiourea
Specifications
Specifications
| Chemical Name or Material | 1-(5-Bromoquinoxalin-6-yl)thiourea |
| CAS | 842138-74-3 |
| Analyte or Component Names | 1-(5-Bromoquinoxalin-6-yl)thiourea |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H7 Br N4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-(5-Bromo-6-quinoxalinyl)thiourea, Thiourea, (5-bromo-6-quinoxalinyl)- (9CI), 5-Bromoquinazolin-6-ylthiourea, Brimonidine Related Compound B, Brimonidine Tartrate Impurity D, Brimonidine Impurity D |
| InChI Formula | InChI=1S/C9H7BrN4S/c10-7-5(14-9(11)15)1-2-6-8(7)13-4-3-12-6/h1-4H,(H3,11,14,15) |
| SMILES | NC(=S)Nc1ccc2nccnc2c1Br |
| IUPAC Name | (5-bromoquinoxalin-6-yl)thiourea |
| Show More |