Learn More
5-Bromoquinazolin-6-ylthiourea, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(5-Bromoquinoxalin-6-yl)thiourea |
| CAS | 842138-74-3 |
| Molecular Formula | C9 H7 Br N4 S |
| Quantity | 1 g |
| Synonym | N-(5-Bromo-6-quinoxalinyl)thiourea, Thiourea, (5-bromo-6-quinoxalinyl)- (9CI), 5-Bromoquinazolin-6-ylthiourea |
| InChI Formula | InChI=1S/C9H7BrN4S/c10-7-5(14-9(11)15)1-2-6-8(7)13-4-3-12-6/h1-4H,(H3,11,14,15) |
| SMILES | NC(=S)Nc1ccc2nccnc2c1Br |
| IUPAC Name | (5-bromoquinoxalin-6-yl)thiourea |
| Molecular Weight (g/mol) | 283.15 |
| Formula Weight | 281.9575 |