CAS RN 76497-39-7
IUPAC Name:
(2S)-3-(acetylsulfanyl)-2-methylpropanoate
Synonyms:
(2S)-3-(acetylsulfanyl)-2-methylpropanoate
Molecular Weight (g/mol):
161.2
Molecular Formula:
C6H9O3S
InChi Key:
VFVHNRJEYQGRGE-SCSAIBSYSA-M
SMILES:
C[C@H](CSC(C)=O)C([O-])=O
1
–
3
of
3
results
1
(S)-(-)-3-Acetylthio-2-methylpropionic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2S)-3-(Acetylthio)-2-methylpropanoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
(S)-(-)-3-Acetylthio-2-methylpropionic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.