Learn More
(S)-(-)-3-Acetylthio-2-methylpropionic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (2S)-3-(Acetylthio)-2-methylpropanoic Acid |
| CAS | 76497-39-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6 H10 O3 S |
| Quantity | 250 mg |
| Synonym | (2S)-3-(Acetylthio)-2-methylpropanoic Acid, Captopril Imp. K (Pharmeuropa) |
| InChI Formula | InChI=1S/C6H10O3S/c1-4(6(8)9)3-10-5(2)7/h4H,3H2,1-2H3,(H,8,9)/t4-/m1/s1 |
| SMILES | C[C@H](CSC(=O)C)C(=O)O |
| IUPAC Name | (2S)-3-acetylsulfanyl-2-methylpropanoic acid |
| Molecular Weight (g/mol) | 162.21 |
| Show More |