Learn More
(2S)-3-(Acetylthio)-2-methylpropanoic Acid, Mikromol™
Product Code. 15315236
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15315236 100 mg 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15315236 Supplier Mikromol Supplier No. MM0067.04
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2S)-3-(Acetylthio)-2-methylpropanoic Acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2S)-3-(Acetylthio)-2-methylpropanoic Acid
CAS 76497-39-7
Analyte or Component Names (2S)-3-(Acetylthio)-2-methylpropanoic Acid
Shipping Condition Room Temperature
Molecular Formula C6 H10 O3 S
Certifications/Compliance ISO 9001
Synonym (2S)-3-(Acetylthio)-2-methylpropanoic Acid, Captopril Imp. K (Pharmeuropa), Captopril Impurity G as (S)-enantiomer
InChI Formula InChI=1S/C6H10O3S/c1-4(6(8)9)3-10-5(2)7/h4H,3H2,1-2H3,(H,8,9)/t4-/m1/s1
SMILES C[C@H](CSC(=O)C)C(=O)O
IUPAC Name (2S)-3-acetylsulfanyl-2-methylpropanoic acid
Molecular Weight (g/mol) 162.21
Quantity 100 mg
Formula Weight 162.0351 g/mol