CAS RN 68751-57-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
[2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl]methanol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolane-4-methanol (Mixture of diastereomers), TRC
CAS: 68751-57-5 Molecular Formula: C11 H11 Br Cl2 O3 Molecular Weight (g/mol): 342.01 Synonym: 1,3-Dioxolane-4-methanol, 2-(bromomethyl)-2-(2,4-dichlorophenyl)-,2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolane-4-methanol,[2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl]methanol IUPAC Name: [2-(bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl]methanol SMILES: OCC1COC(CBr)(O1)c2ccc(Cl)cc2Cl