Learn More
2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolane-4-methanol (Mixture of diastereomers), TRC
Product Code. 17993285
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993285 100 mg 100mg
17903485 1 g 1g
17983285 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17993285 Supplier Toronto Research Chemicals Supplier No. TRCB685525100MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 68751-57-5
Molecular Formula C11 H11 Br Cl2 O3
Molecular Weight (g/mol) 342.01
Synonym 1,3-Dioxolane-4-methanol, 2-(bromomethyl)-2-(2,4-dichlorophenyl)-, 2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolane-4-methanol, [2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl]methanol
IUPAC Name [2-(bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl]methanol
SMILES OCC1COC(CBr)(O1)c2ccc(Cl)cc2Cl
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C11H11BrCl2O3/c12-6-11(16-5-8(4-15)17-11)9-2-1-7(13)3-10(9)14/h1-3,8,15H,4-6H2
Formula Weight 339.9269
Chemical Name or Material 2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolane-4-methanol (Mixture of diastereomers)