Learn More
[2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl]methanol, Mikromol™
Description
[2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl]methanol
Specifications
Specifications
| Chemical Name or Material | 2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolane-4-methanol (Mixture of diastereomers) |
| CAS | 68751-57-5 |
| Analyte or Component Names | 2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolane-4-methanol (Mixture of diastereomers) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H11 Br Cl2 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1,3-Dioxolane-4-methanol, 2-(bromomethyl)-2-(2,4-dichlorophenyl)-, 2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolane-4-methanol, [2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl]methanol |
| InChI Formula | InChI=1S/C11H11BrCl2O3/c12-6-11(16-5-8(4-15)17-11)9-2-1-7(13)3-10(9)14/h1-3,8,15H,4-6H2 |
| SMILES | OCC1COC(CBr)(O1)c2ccc(Cl)cc2Cl |
| IUPAC Name | [2-(bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl]methanol |
| Show More |