accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 51755-66-9

H3COHSCH3

CAS RN 51755-66-9

IUPAC Name: 3-(methylsulfanyl)hexan-1-ol
Synonyms: 3-(methylsulfanyl)hexan-1-ol
Molecular Weight (g/mol): 148.26
Molecular Formula: C7H16OS
InChi Key: JSASXSHMJYRPCM-UHFFFAOYNA-N
SMILES: CCCC(CCO)SC
Molecular Weight (g/mol) 
  • (3)
  • (1)
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results