Learn More
3-(Methylthio)hexanol, 99%, Thermo Scientific™
Product Code. 11490427
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490427 25 g 25g
11480427 5 g 5g
11400437 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11490427 Supplier Thermo Scientific Alfa Aesar Supplier No. B21786.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51755-66-9
Molecular Formula C7H16OS
Molecular Weight (g/mol) 148.26
MDL Number MFCD00010674
InChI Key JSASXSHMJYRPCM-UHFFFAOYNA-N
Synonym 3-methylthio-1-hexanol, 3-methylthio hexanol, 1-hexanol, 3-methylthio, 3-methylthio hexan-1-ol, 3-methylthiohexanol, 3-methylsulfanyl hexan-1-ol, r-3-methylthio-1-hexanol, 3-methylmercapto-1-hexanol, fema no. 3438
PubChem CID 65413
SMILES CCCC(CCO)SC
Specifications
Density 0.966
Boiling Point 61°C to 62°C (10 mmHg)
Flash Point 107°C (226°F)
Linear Formula CH3CH2CH2CH(SCH3)CH2CH2OH
Refractive Index 1.4759
Quantity 25 g
UN Number UN3334
IUPAC Name 3-methylsulfanylhexan-1-ol
Formula Weight 148.26
Percent Purity 99%
Chemical Name or Material 3-(Methylthio)hexanol