Learn More
3-(Methylthio)hexan-1-ol, Dr. Ehrenstorfer
Product Code. 30326797
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326797 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30326797 Supplier Dr. Ehrenstorfer Supplier No. DREC15144210
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-(Methylthio)hexan-1-ol
CAS 51755-66-9
Analyte or Component Names 3-(Methylthio)hexan-1-ol
Shipping Condition +20°C
Molecular Formula C7 H16 O S
Certifications/Compliance ISO/IEC 17025
Synonym 3-(Methylthio)-1-hexanol, 3-(Methylthio)hexanol, 1-Hexanol, 3-(methylthio)-
InChI Formula InChI=1S/C7H16OS/c1-3-4-7(9-2)5-6-8/h7-8H,3-6H2,1-2H3
SMILES CCCC(CCO)SC
IUPAC Name 3-methylsulfanylhexan-1-ol
Molecular Weight (g/mol) 148.2663
Quantity 250 mg
Formula Weight 148.0922 g/mol