CAS RN 43109-77-9
CAS RN 43109-77-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
o-Adamantylanisole, TRC
CAS: 43109-77-9 Molecular Formula: C17 H22 O Molecular Weight (g/mol): 242.36 Synonym: 1-(2-Methoxyphenyl)tricyclo[3.3.1.13,7]decane,Adapalene Imp. C (EP) IUPAC Name: 1-(2-methoxyphenyl)adamantane SMILES: COc1ccccc1C23CC4CC(CC(C4)C2)C3
2
1-(2-Methoxyphenyl)tricyclo[3.3.1.13,7]decane, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.