Learn More
o-Adamantylanisole, TRC
Product Code. 30138277
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138277 500 mg 500mg
30137329 250 mg 250mg
30011800 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138277 Supplier Toronto Research Chemicals Supplier No. TRCA208250500MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 43109-77-9
Molecular Formula C17 H22 O
Molecular Weight (g/mol) 242.36
Synonym 1-(2-Methoxyphenyl)tricyclo[3.3.1.13,7]decane, Adapalene Imp. C (EP)
IUPAC Name 1-(2-methoxyphenyl)adamantane
SMILES COc1ccccc1C23CC4CC(CC(C4)C2)C3
Specifications
Analyte or Component Names 1-(2-Methoxyphenyl)tricyclo[3.3.1.13,7]decane
Shipping Condition Room Temperature
InChI Formula InChI=1S/C17H22O/c1-18-16-5-3-2-4-15(16)17-9-12-6-13(10-17)8-14(7-12)11-17/h2-5,12-14H,6-11H2,1H3
Solution Type Neat
Industry Type Pharmaceutical
Quantity 500 mg
Formula Weight 242.1671
Percent Purity >95%
Chemical Name or Material 1-(2-Methoxyphenyl)tricyclo[3.3.1.13,7]decane
Grade HPLC