CAS RN 391860-73-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
6-Chloro-2-cyclopropyl-4-(trifluoromethyl)-quinoline, TRC
CAS: 391860-73-4 Molecular Formula: C13 H9 Cl F3 N Molecular Weight (g/mol): 271.67 Synonym: 6-Chloro-2-cyclopropyl-4-(trifluoromethyl)quinoline,Efavirenz Related Compound C (USP) IUPAC Name: 6-chloro-2-cyclopropyl-4-(trifluoromethyl)quinoline SMILES: FC(F)(F)c1cc(nc2ccc(Cl)cc12)C3CC3
2
Efavirenz USP Related Compound C, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.