Learn More
6-Chloro-2-cyclopropyl-4-(trifluoromethyl)-quinoline, TRC
Product Code. 30367892
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367892 1 g 1g
30366922 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30367892 Supplier Toronto Research Chemicals Supplier No. TRCC3655451G
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 391860-73-4
Molecular Formula C13 H9 Cl F3 N
Molecular Weight (g/mol) 271.67
Synonym 6-Chloro-2-cyclopropyl-4-(trifluoromethyl)quinoline, Efavirenz Related Compound C (USP)
IUPAC Name 6-chloro-2-cyclopropyl-4-(trifluoromethyl)quinoline
SMILES FC(F)(F)c1cc(nc2ccc(Cl)cc12)C3CC3
Specifications
Quantity 1 g
InChI Formula InChI=1S/C13H9ClF3N/c14-8-3-4-11-9(5-8)10(13(15,16)17)6-12(18-11)7-1-2-7/h3-7H,1-2H2
Formula Weight 271.0376
Chemical Name or Material 6-chloro-2-cyclopropyl-4-(trifluoromethyl)quinoline