Learn More
Efavirenz USP Related Compound C, Mikromol™
Product Code. 30462357
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462357 4 x 25 mg 100mg
30050748 25 mg 25mg
30049397 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30462357 Supplier Mikromol Supplier No. MM0919.0500254AMP
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Efavirenz USP Related Compound C, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-chloro-2-cyclopropyl-4-(trifluoromethyl)quinoline
CAS 391860-73-4
Analyte or Component Names 6-chloro-2-cyclopropyl-4-(trifluoromethyl)quinoline
Shipping Condition Room Temperature
Molecular Formula C13 H9 Cl F3 N
Certifications/Compliance ISO 17025
Synonym 6-Chloro-2-cyclopropyl-4-(trifluoromethyl)quinoline, Efavirenz Related Compound C (USP), Efavirenz Related Compound C
InChI Formula InChI=1S/C13H9ClF3N/c14-8-3-4-11-9(5-8)10(13(15,16)17)6-12(18-11)7-1-2-7/h3-7H,1-2H2
Solution Type Neat
SMILES FC(F)(F)c1cc(nc2ccc(Cl)cc12)C3CC3
IUPAC Name 6-chloro-2-cyclopropyl-4-(trifluoromethyl)quinoline
Molecular Weight (g/mol) 271.67
Quantity 4 x 25 mg
Formula Weight 271.0376 g/mol