accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 328-90-5

HOOHOFFF

CAS RN 328-90-5

IUPAC Name: 2-hydroxy-4-(trifluoromethyl)benzoic acid
Synonyms: 2-hydroxy-4-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 206.12
Molecular Formula: C8H5F3O3
InChi Key: XMLFPUBZFSJWCN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results