Learn More
Desacetyl Triflusal, TRC
Specifications
Specifications
| Chemical Name or Material | Triflusal impurity B |
| CAS | 328-90-5 |
| Molecular Formula | C8H5F3O3 |
| Quantity | 100 mg |
| Synonym | Triflusal Imp. B (EP) |
| InChI Formula | InChI=1S/C8H5F3O3/c9-8(10,11)4-1-2-5(7(13)14)6(12)3-4/h1-3,12H,(H,13,14) |
| SMILES | OC(=O)c1ccc(cc1O)C(F)(F)F |
| Molecular Weight (g/mol) | 206.12 |