Learn More
Triflusal EP Impurity B, Mikromol™
Product Code. 30464063
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464063 4 x 25 mg 100mg
30051355 100 mg 100mg
30052659 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30464063 Supplier Mikromol Supplier No. MM3414.0100254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Triflusal EP Impurity B, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(Trifluoromethyl)salicylic Acid
CAS 328-90-5
Analyte or Component Names 4-(Trifluoromethyl)salicylic Acid
Shipping Condition Room Temperature
Molecular Formula C8 H5 F3 O3
Certifications/Compliance ISO 17025
Synonym 2-Hydroxy-4-(trifluoromethyl)benzoic Acid (4-(Trifluoromethyl)salicylic Acid), Triflusal Imp. B (EP), 2-Hydroxy-4-(trifluoromethyl)-benzoic Acid, 2-Hydroxy-4-(trifluoromethyl)benzoic acid, 4-(Trifluoromethyl)salicylic acid, 4-Trifluoromethyl-2-hydroxybenzoic acid, p-(Trifluoromethyl)salicylic acid, 2,4-Cresotic acid, alpha,alpha,alpha-trifluoro- (6CI,7CI,8CI), Triflusal Impurity B
InChI Formula InChI=1S/C8H5F3O3/c9-8(10,11)4-1-2-5(7(13)14)6(12)3-4/h1-3,12H,(H,13,14)
Solution Type Neat
SMILES OC(=O)c1ccc(cc1O)C(F)(F)F
IUPAC Name 2-hydroxy-4-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol) 206.12
Quantity 4 x 25 mg
Formula Weight 206.0191 g/mol