accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3254-89-5

HClHON

CAS RN 3254-89-5

IUPAC Name: hydrogen 1,1-diphenyl-4-(piperidin-1-yl)butan-1-ol chloride
Synonyms: hydrogen diphenidol chloride
Molecular Weight (g/mol): 345.91
Molecular Formula: C21H28ClNO
InChi Key: AVZIYZHXZAYGJS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OC(CCCN1CCCCC1)(C1=CC=CC=C1)C1=CC=CC=C1
Certifications/Compliance 
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results