Learn More
Diphenidol hydrochloride, MedChemExpress
Product Code. 16428026
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16428026 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16428026 Supplier Medchem Express Supplier No. HYA0082
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Diphenidol hydrochloride (Difenidol hydrochloride) is a non-selective muscarinic M1-M4 receptor antagonist, has anti-arrhythmic activity. Diphenidol hydrochloride is also a potent non-specific blocker of voltage-gated ion channels (Na+, K+, and Ca2+) in neuronal cells.
Diphenidol hydrochloride (Difenidol hydrochloride) is a non-selective muscarinic M1-M4 receptor antagonist, has anti-arrhythmic activity. Diphenidol hydrochloride is also a potent non-specific blocker of voltage-gated ion channels (Na+, K+, and Ca2+) in neuronal cells.
Specifications
Chemical Name or Material Diphenidol hydrochloride
Physical Form Solid
CAS 3254-89-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H28ClNO
For Use With (Application) Neuroscience-Neuromodulation
Synonym Difenidol hydrochloride
Solubility Information DMSO : ≥ 46 mg/mL (132.98 mM) ∣H2O : 16.67 mg/mL (48.19 mM; Need ultrasonic)
SMILES OC(C1=CC=CC=C1)(C2=CC=CC=C2)CCCN3CCCCC3.[H]Cl
Molecular Weight (g/mol) 345.91
Formula Weight 345.91
Percent Purity 99.0%
Grade Research