Learn More
Difenidol Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Difenidol Hydrochloride |
| CAS | 3254-89-5 |
| Analyte or Component Names | Difenidol Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H27 N O . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ansmin, Cefadol, Celmidol, Difenidol hydrochloride, Difenidolin, Diphenidol hydrochloride, Maniol, Mecalmin, Pineroro, SKF 478A, Satanolon, Tenesdol, Vontrol, Wansar, 1-Piperidinebutanol, alpha,alpha-diphenyl-, hydrochloride (1:1), 1-Piperidinebutanol, alpha,alpha-diphenyl-, hydrochloride (6CI,7CI,8CI,9CI), SKF-478A |
| InChI Formula | InChI=1S/C21H27NO.ClH/c23-21(19-11-4-1-5-12-19,20-13-6-2-7-14-20)15-10-18-22-16-8-3-9-17-22;/h1-2,4-7,11-14,23H,3,8-10,15-18H2;1H |
| Solution Type | Neat |
| SMILES | Cl.OC(CCCN1CCCCC1)(c2ccccc2)c3ccccc3 |
| Show More |