accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3244-90-4

CH3OPSOCH3OPSOCH3OCH3

CAS RN 3244-90-4

IUPAC Name: O,O-dipropyl {[dipropoxy(sulfanylidene)-λ⁵-phosphanyl]oxy}phosphonothioate
Synonyms: tetrapropyl thiopyrophosphorate
Molecular Weight (g/mol): 378.42
Molecular Formula: C12H28O5P2S2
InChi Key: IIDFEIDMIKSJSV-UHFFFAOYSA-N
SMILES: CCCOP(=S)(OCCC)OP(=S)(OCCC)OCCC
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results