Learn More
EPA Method 8141 Organophosphorus Pesticide Mixture 429 200 μg/mL in n-Hexane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Aspon∣Chlorpyrifos-methyl∣Crotoxyphos∣Dichlofenthion∣Dicrotophos∣Fenitrothion∣Fonofos∣Thionazin∣Trichlorphon |
| CAS | 3244-90-4, 5598-13-0, 7700-17-6, 97-17-6, 141-66-2, 122-14-5 |
| Analyte or Component Names | Aspon∣Chlorpyrifos-methyl∣Crotoxyphos∣Dichlofenthion∣Dicrotophos∣Fenitrothion∣Fonofos∣Thionazin∣Trichlorphon |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Hexane |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Aspon 200 μg/mL∣Chlorpyrifos-methyl 200 μg/mL∣Crotoxyphos 200 μg/mL∣Dichlofenthion 200 μg/mL∣Dicrotophos 200 μg/mL∣Fenitrothion 200 μg/mL∣Fonofos 200 μg/mL∣Thionazin 200 μg/mL∣Trichlorphon 200 μg/mL |