accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3221-61-2

H3CCH3CH3

CAS RN 3221-61-2

IUPAC Name: 2-methyloctane
Synonyms: 2-methyloctane
Molecular Weight (g/mol): 128.26
Molecular Formula: C9H20
InChi Key: ZUBZATZOEPUUQF-UHFFFAOYSA-N
SMILES: CCCCCCC(C)C
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results