Learn More
2-Methyloctane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2-Methyloctane |
| CAS | 3221-61-2 |
| Analyte or Component Names | 2-Methyloctane |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H20 |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | 2-Methyloctane, NSC 23695 |
| InChI Formula | InChI=1S/C9H20/c1-4-5-6-7-8-9(2)3/h9H,4-8H2,1-3H3 |
| Solution Type | Neat |
| SMILES | CCCCCCC(C)C |
| Show More |