CAS RN 31857-31-5
CAS RN 31857-31-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
2-Cyano-1,3-dimethylguanidine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
N-Cyano-N’,N’-dimethyl-guanidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Cyano-N’,N' '-dimethyl-guanidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
N-Cyano-N’,N' '-dimethyl-guanidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.