Learn More
N-Cyano-N’,N’-dimethyl-guanidine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Cyano-1,3-dimethylguanidine |
| CAS | 31857-31-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C4 H8 N4 |
| Quantity | 100 mg |
| Synonym | 2-Cyano-1,3-dimethylguanidine, Cimetidine Hydrochloride Imp. G (EP) |
| InChI Formula | InChI=1S/C4H8N4/c1-6-4(7-2)8-3-5/h1-2H3,(H2,6,7,8) |
| SMILES | CNC(=NC#N)NC |
| IUPAC Name | 2-cyano-1,3-dimethylguanidine |
| Molecular Weight (g/mol) | 112.13 |
| Show More |