Learn More
2-Cyano-1,3-dimethylguanidine, Mikromol™
Description
2-Cyano-1,3-dimethylguanidine
Specifications
Specifications
| Chemical Name or Material | 2-Cyano-1,3-dimethylguanidine |
| CAS | 31857-31-5 |
| Analyte or Component Names | 2-Cyano-1,3-dimethylguanidine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C4 H8 N4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Cimetidine Hydrochloride Imp. G (EP), Cimetidine Imp. G (EP), 2-Cyano-1,3-dimethylguanidine, Cimetidine Hydrochloride Impurity G, Cimetidine Impurity G, Cimetidine Impurity G |
| InChI Formula | InChI=1S/C4H8N4/c1-6-4(7-2)8-3-5/h1-2H3,(H2,6,7,8) |
| SMILES | CNC(=NC#N)NC |
| IUPAC Name | 2-cyano-1,3-dimethylguanidine |
| Show More |